missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dithiobis(2-nitrobenzoic acid), 99%

Product Code. p-7021834
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11476887

Brand: Thermo Scientific Alfa Aesar A14331.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol

Reagent for determination of sufhydryl groups.5,5'-dithiobis-(2-nitrobenzoic acid) is widely utilized as a non-fluorescent probe to quantify the number or the concentration of thiol groups in a sample. It acts as a cleavable cross-linking agent. It finds an important application as a sensitive reagent for determining free sulfhydryl content in proteins, peptides, and tissues. It is also employed in high-performance liquid chromatography (HPLC) to determine the alkylthiol content.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for determination of sufhydryl groups.5,5′-dithiobis-(2-nitrobenzoic acid) is widely utilized as a non-fluorescent probe to quantify the number or the concentration of thiol groups in a sample. It acts as a cleavable cross-linking agent. It finds an important application as a sensitive reagent for determining free sulfhydryl content in proteins, peptides, and tissues. It is also employed in high-performance liquid chromatography (HPLC) to determine the alkylthiol content.

Solubility
Soluble in water and ethanol.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-78-3
Molecular Formula C14H8N2O8S2
Molecular Weight (g/mol) 396.34
MDL Number MFCD00007140
InChI Key KIUMMUBSPKGMOY-UHFFFAOYSA-N
Synonym dtnb, 5,5'-dithiobis 2-nitrobenzoic acid, ellman's reagent, 3-carboxy-4-nitrophenyl disulfide, dithionitrobenzoic acid, dithiobisnitrobenzoic acid, 5,5'-disulfanediylbis 2-nitrobenzoic acid, benzoic acid, 3,3'-dithiobis 6-nitro, 3,3'-dithiobis 6-nitrobenzoic acid, 5,5'-dithio-bis 2-nitrobenzoic acid
PubChem CID 6254
ChEBI CHEBI:86228
IUPAC Name 5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
SMILES OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 1 g
Beilstein 1896821
Solubility Information Soluble in water and ethanol.
Formula Weight 396.36
Percent Purity 99%
Chemical Name or Material 5,5'-Dithiobis(2-nitrobenzoic acid)
compliance-icons
Product Identifier
  • 5,5'-Dithiobis(2-nitrobenzoic acid)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 5,5'-Dithiobis(2-nitrobenzoic acid)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.