missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin-3-butyric Acid, TRC

Product Code. 30380774
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380774 5 mg
30372673 10 mg
30377632 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380774 Supplier Toronto Research Chemicals Supplier No. TRCD4916805MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56976-66-0
Molecular Formula C19 H18 N2 O4
Molecular Weight (g/mol) 338.36
IUPAC Name 4-(2,5-dioxo-4,4-diphenylimidazolidin-1-yl)butanoic acid
SMILES OC(=O)CCCN1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

Tekniske data

Quantity 5 mg
InChI Formula InChI=1S/C19H18N2O4/c22-16(23)12-7-13-21-17(24)19(20-18(21)25,14-8-3-1-4-9-14)15-10-5-2-6-11-15/h1-6,8-11H,7,12-13H2,(H,20,25)(H,22,23)
Formula Weight 338.1267
Chemical Name or Material 5,5-Diphenylhydantoin-3-butyric Acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.