missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin-3-butyric Acid, TRC

Product Code. 30372673
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372673 10 mg
30377632 100 mg
30380774 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372673 Supplier Toronto Research Chemicals Supplier No. TRCD49168010MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56976-66-0
Molecular Formula C19 H18 N2 O4
Molecular Weight (g/mol) 338.36
IUPAC Name 4-(2,5-dioxo-4,4-diphenylimidazolidin-1-yl)butanoic acid
SMILES OC(=O)CCCN1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H18N2O4/c22-16(23)12-7-13-21-17(24)19(20-18(21)25,14-8-3-1-4-9-14)15-10-5-2-6-11-15/h1-6,8-11H,7,12-13H2,(H,20,25)(H,22,23)
Formula Weight 338.1267
Chemical Name or Material 5,5-Diphenylhydantoin-3-butyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.