missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Diphenyl Hydantoin Sodium, TRC
Specifications
Specifications
| Chemical Name or Material | Phenytoin sodium |
| CAS | 630-93-3 |
| Molecular Formula | C15 H11 N2 O2 . Na |
| Quantity | 1 g |
| Synonym | Phenytoin Sodium, 5,5-Diphenyl-2,4-imidazolidinedione sodium salt (1:1), 5,5-Diphenyl-2,4-imidazolidinedione monosodium salt, 5,5-Diphenylhydantoin sodium salt, 5,5-Diphenylhydantoin sodium, Aleviatin sodium, Antisacer, Danten, Difenin, Difhydan, Dilantin, Diphantoine, Diphenin, Diphenine, Diphenylan sodium, Diphenylhydantoin sodium, Ditoin, Enkefal, Epanutin, Epelin, Epilan D, Epsolin, Eptoin, Fenitoin sodium, Hydantin, Hydantoinal, M-toin, Minetoin, Phenyloin, Phenytoin sodium, Phenytoin soluble, Prompt, Sodium 5,5-diphenyl-2,4-imidazolidinedione, Sodium 5,5-diphenylhydantoin, Sodium diphenylhydantoin, Sodium diphenylhydantoinate, Sodium phenytoin, Solantyl, Soluble Phenytoin, Tacosal |
| InChI Formula | InChI=1S/C15H12N2O2.Na/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,(H2,16,17,18,19);/q;+1/p-1 |
| SMILES | [Na+].[O-]C1=NC(=O)C(N1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | sodium;4-oxo-5,5-diphenyl-1H-imidazol-2-olate |
| Molecular Weight (g/mol) | 274.25 |
| Formula Weight | 274.0718 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?