missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethylhydantoin, 97%

Product Code. 11312136
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312136 100 g 100g
11322136 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312136 Supplier Thermo Scientific Alfa Aesar Supplier No. L03388.22
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-71-4 | C5H8N2O2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-71-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005266
InChI Key YIROYDNZEPTFOL-UHFFFAOYSA-N
Synonym 5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin
PubChem CID 6491
IUPAC Name 5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)NC(=O)NC1=O

Specifications

Melting Point 174°C to 178°C
Flash Point 193°C (379°F)
Odor Odorless
Quantity 100 g
Beilstein 2827
Formula Weight 128.13
Percent Purity 97%
Chemical Name or Material 5,5-Dimethylhydantoin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.