missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Chloro-2-nitrophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15400796
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1449144-52-8
C13H9BClFN2O5
338.482
MFCD20040135
YARGFVSGOJRMQM-UHFFFAOYSA-N
5-5-chloro-2-nitrophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-5-chloro-2-nitrophenyl carbamoyl-2-fluorophenylboronic acid
73995641
[5-[(5-chloro-2-nitrophenyl)carbamoyl]-2-fluorophenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)[N+](=O)[O-])F)(O)O
This item is not returnable. View return policy
1449144-52-8
C13H9BClFN2O5
338.482
MFCD20040135
YARGFVSGOJRMQM-UHFFFAOYSA-N
5-5-chloro-2-nitrophenylcarbamoyl-2-fluorobenzeneboronic acid, 5-5-chloro-2-nitrophenyl carbamoyl-2-fluorophenylboronic acid
73995641
[5-[(5-chloro-2-nitrophenyl)carbamoyl]-2-fluorophenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)[N+](=O)[O-])F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 338.49
Percent Purity 97%
Chemical Name or Material 5-(5-Chloro-2-nitrophenylcarbamoyl)-2-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'