missing translation for 'onlineSavingsMsg'
Learn More

5,5-bis(2-Fluorophenyl)-5-hydroxyvaleric Acid, TRC

Product Code. 30402389
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402389 1 g 1g
30399411 500 mg 500mg
30404668 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402389 Supplier Toronto Research Chemicals Supplier No. TRCF4896151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,5-bis(2-Fluorophenyl)-5-hydroxyvaleric Acid
Analyte or Component Names 5,5-bis(2-Fluorophenyl)-5-hydroxyvaleric Acid
Shipping Condition Room Temperature
Molecular Formula C17 H16 F2 O3
Synonym 5,5-bis(2-Fluorophenyl)-5-hydroxypentanoic Acid
InChI Formula InChI=1S/C17H16F2O3/c18-14-8-3-1-6-12(14)17(22,11-5-10-16(20)21)13-7-2-4-9-15(13)19/h1-4,6-9,22H,5,10-11H2,(H,20,21)
Solution Type Neat
SMILES OC(=O)CCCC(O)(c1ccccc1F)c2ccccc2F
IUPAC Name 5,5-bis(2-fluorophenyl)-5-hydroxypentanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 306.304
Quantity 1 g
Formula Weight 306.107
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.