missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-bis(2-Fluorophenyl)-5-hydroxyvaleric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5,5-bis(2-Fluorophenyl)-5-hydroxyvaleric Acid |
| Analyte or Component Names | 5,5-bis(2-Fluorophenyl)-5-hydroxyvaleric Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H16 F2 O3 |
| Synonym | 5,5-bis(2-Fluorophenyl)-5-hydroxypentanoic Acid |
| InChI Formula | InChI=1S/C17H16F2O3/c18-14-8-3-1-6-12(14)17(22,11-5-10-16(20)21)13-7-2-4-9-15(13)19/h1-4,6-9,22H,5,10-11H2,(H,20,21) |
| Solution Type | Neat |
| SMILES | OC(=O)CCCC(O)(c1ccccc1F)c2ccccc2F |
| IUPAC Name | 5,5-bis(2-fluorophenyl)-5-hydroxypentanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?