missing translation for 'onlineSavingsMsg'
Learn More

5,5,8,8-Tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylic acid, 97%, Thermo Scientific™

Product Code. p-3075742
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10066012 250 mg Amber Glass Bottle 250mg
10680135 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10066012 Supplier Thermo Scientific Maybridge Supplier No. CC27101CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 103031-30-7
Molecular Formula C15H20O2
Molecular Weight (g/mol) 232.323
MDL Number MFCD04115382
InChI Key KSEZYZSBKCPEKP-UHFFFAOYSA-N
Synonym 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl, 2-naphthalenecarboxylicacid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl, acmc-1c8hs, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthoic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthoic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylicacid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-naphthalene-2-carboxylic acid
PubChem CID 227638
IUPAC Name 5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-carboxylic acid
SMILES CC1(CCC(C2=C1C=CC(=C2)C(=O)O)(C)C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 232.32
Percent Purity 97%
Chemical Name or Material 5,5,8,8-Tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.