missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxyphenyl)pentanoic acid, 98%, Thermo Scientific™

Product Code. 11486137
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486137 25 g 25g
11476137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486137 Supplier Thermo Scientific Alfa Aesar Supplier No. B25107.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-(4-Hydroxyphenyl)pentanoic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4654-08-4

Specifications

Melting Point 116°C to 120°C
Molecular Formula C7H4N2O2
MDL Number MFCD00013439
Quantity 25 g
Synonym 3,4-pyridinedicarboximide, cinchomeronimide, 3,4-pyridinecarboximide, 1h-pyrrolo 3,4-c pyridine-1,3 2h-dione, 2h-pyrrolo 3,4-c pyridine-1,3-dione, pyrrolo 3,4-c pyridine-1,3-dione, cinchomeronic acid imide, 4-azaphthalimid, pyridine-3,4-dicarboximide, 1h,2h,3h-pyrrolo 3,4-c pyridine-1,3-dione
Solubility Information Soluble in water.
InChI Key SJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILES O=C1NC(=O)C2=CN=CC=C12
IUPAC Name pyrrolo[3,4-c]pyridine-1,3-dione
Molecular Weight (g/mol) 148.12
PubChem CID 72926
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 5-(4-Hydroxyphenyl)pentanoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.