missing translation for 'onlineSavingsMsg'
Learn More

5-[(4-Hydroxyphenyl)methyl]oxolan-2-one, TRC

Product Code. 30391990
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391990 10 mg -
30406016 50 mg 50mg
30400048 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391990 Supplier Toronto Research Chemicals Supplier No. TRCH10069310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871329-30-5
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
IUPAC Name 5-[(4-hydroxyphenyl)methyl]oxolan-2-one
SMILES Oc1ccc(CC2CCC(=O)O2)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12O3/c12-9-3-1-8(2-4-9)7-10-5-6-11(13)14-10/h1-4,10,12H,5-7H2
Formula Weight 192.0786
Chemical Name or Material 5-[(4-Hydroxyphenyl)methyl]oxolan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.