missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxyphenyl)-5-phenylhydantoin, TRC

Product Code. 30395662
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395662 50 mg
30395276 100 mg
30392212 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395662 Supplier Toronto Research Chemicals Supplier No. TRCH51677050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2784-27-2
Molecular Formula C15 H12 N2 O3
Molecular Weight (g/mol) 268.2674
Synonym Hydantoin, 5-(p-hydroxyphenyl)-5-phenyl-, (p-Hydroxyphenyl)phenylhydantoin, (±)-5-(4-Hydroxyphenyl-5-phenylhydantoin, 4-Hydroxydiphenylhydantoin, 4-Hydroxyphenytoin, 5-(4-Hydroxyphenyl)-5-phenylhydantoin, 5-(4'-Hydroxyphenyl)-5-phenylhydantoin, 5-(p-Hydroxyphenyl)-5-phenylhydantoin, 5-Phenyl-5-p-hydroxyphenylhydantoin, Hydroxydiphenylhydantoin, NSC 156081, dl-5-(p-Hydroxyphenyl-5-phenylhydantoin, p-Hydroxydiphenylhydantoin, p-Hydroxyphenytoin, 2,4-Imidazolidinedione, 5-(4-hydroxyphenyl)-5-phenyl-
IUPAC Name 5-(4-hydroxyphenyl)-5-phenylimidazolidine-2,4-dione
SMILES Oc1ccc(cc1)C2(NC(=O)NC2=O)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H12N2O3/c18-12-8-6-11(7-9-12)15(10-4-2-1-3-5-10)13(19)16-14(20)17-15/h1-9,18H,(H2,16,17,19,20)
Formula Weight 268.0848
Chemical Name or Material 5-(4-Hydroxyphenyl)-5-phenylhydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.