missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxybenzyl)thiazolidine-2,4-dione, TRC

Product Code. 30398299
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398299 1 g -
30395914 100 mg -
30403989 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398299 Supplier Toronto Research Chemicals Supplier No. TRCH8299801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74772-78-4
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
Synonym 5-[(4-Hydroxyphenyl)methyl]-2,4-thiazolidinedione, U 90441
IUPAC Name 5-[(4-hydroxyphenyl)methyl]-1,3-thiazolidine-2,4-dione
SMILES Oc1ccc(CC2SC(=O)NC2=O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H9NO3S/c12-7-3-1-6(2-4-7)5-8-9(13)11-10(14)15-8/h1-4,8,12H,5H2,(H,11,13,14)
Formula Weight 223.0303
Chemical Name or Material 5-(4-Hydroxybenzyl)thiazolidine-2,4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.