missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)valeric acid, 97%

Product Code. 11324835
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324835 250 mg 250mg
11304835 1 g 1g
11314835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324835 Supplier Thermo Scientific Alfa Aesar Supplier No. L17935.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24484-22-8 | C11H12FO2 | 195.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24484-22-8
Molecular Formula C11H12FO2
Molecular Weight (g/mol) 195.21
MDL Number MFCD00800242
InChI Key BXEFPLJKYWEEAN-UHFFFAOYSA-M
PubChem CID 2734670
IUPAC Name 5-(4-fluorophenyl)pentanoic acid
SMILES [O-]C(=O)CCCCC1=CC=C(F)C=C1

Specifications

Melting Point 73°C to 75°C
Quantity 250 mg
Beilstein 7918899
Synonym 5-4-fluorophenyl valeric acid, 5-4-fluorophenyl pentanoic acid, benzenepentanoic acid, 4-fluoro, 5-4'-fluorophenyl valeric acid, acmc-1cj6r, benzenepentanoic acid,4-fluoro, 5-4-fluorophenyl-pentanoic acid, 5-4-fluoro-phenyl-pentanoic acid
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 5-(4-Fluorophenyl)valeric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.