missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)cyclohexane-1,3-dione, 95%, Thermo Scientific™

Product Code. 10282782
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10282782 1 g 1g
10029341 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10282782 Supplier Thermo Scientific Maybridge Supplier No. CD11363DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 55579-72-1
Molecular Formula C12H11FO2
Molecular Weight (g/mol) 206.22
MDL Number MFCD00174037
InChI Key LBTLJACXBCUFEF-UHFFFAOYSA-N
Synonym 5-4-fluorophenyl cyclohexane-1,3-dione, 5-4-fluorophenyl-1,3-cyclohexanedione, 5-4-fluoroophenyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 5-4-fluorophenyl, 5-4-fluoro-phenyl-cyclohexane-1,3-dione, acmc-20am8h, 4-3,5-dioxocyclohex-1-yl fluorobenzene, 1,3-cyclohexanedione,5-4-fluorophenyl, 5-4-fluoro-phenyl cyclohexane-1,3-dione, 5-4-fluorophenyl cyclohexane-1, 3-dione
PubChem CID 730136
IUPAC Name 5-(4-fluorophenyl)cyclohexane-1,3-dione
SMILES FC1=CC=C(C=C1)C1CC(=O)CC(=O)C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 206.21
Percent Purity 95%
Chemical Name or Material 5-(4-Fluorophenyl)cyclohexane-1, 3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.