missing translation for 'onlineSavingsMsg'
Learn More

5-[(4-Fluorobenzylidene]-2,4-thiazolidinedione, TRC

Product Code. 30394420
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394420 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394420 Supplier Toronto Research Chemicals Supplier No. TRCF588650500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 262601-87-6
Molecular Formula C10H6FNO2S
Quantity 500 mg
Synonym 5-[(4-Fluorophenyl)methylene]-2,4-thiazolidinedione
InChI Formula InChI=1S/C10H6FNO2S/c11-7-3-1-6(2-4-7)5-8-9(13)12-10(14)15-8/h1-5H,(H,12,13,14)
SMILES Fc1ccc(C=C2SC(=O)NC2=O)cc1
IUPAC Name 5-[(4-fluorophenyl)methylidene]-1,3-thiazolidine-2,4-dione
Molecular Weight (g/mol) 223.22
Formula Weight 223.0103
Chemical Name or Material 5-[4-Fluorobenzylidene]-2,4-thiazolidinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.