missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-(4-Dimethylaminobenzylidene)rhodanine, 99%

Product Code. 10665392 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665392 25 g Glass bottle 25g
10307520 10 g Glass bottle 10g
10574264 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10665392 Supplier Thermo Scientific Chemicals Supplier No. 151470250
  • ONLINE EXCLUSIVE PRICE
    £103.00
    £84.65 / 25g
    Save £18.35
    17% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-(4-Dimethylaminobenzylidene)rhodanine, 99%, Quantity: 100g, Packaging: Glass bottle, Red, Melting Point: 285 deg.C to 288 deg.C, Molecular Weight: 264.35, Percent Purity: 99%, Appearance: (0.3% soln. in acetone) clear, Assay Percent Range: 99%, Beilstein: 27, 433, CAS: 536-17-4 | CAS: 536-17-4 | C12H12N2OS2 | 264.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-17-4
Molecular Formula C12H12N2OS2
Molecular Weight (g/mol) 264.35
MDL Number MFCD00064857
InChI Key JJRVRELEASDUMY-UHFFFAOYSA-N
Synonym unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
PubChem CID 2723826
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 285°C to 288°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 27, 433
Merck Index 15, 3253
Solubility Information (0.03% in acetone) Clear
Formula Weight 264.35
Percent Purity 99%
Physical Form Fluffy Powder
Chemical Name or Material 5-(4-Dimethylaminobenzylidene)rhodanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.