missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)-2-furoic acid, 95+%

Product Code. 10096452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10096452 5 g 5g
10629194 1 g 1g
11926171 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096452 Supplier Thermo Scientific Acros Supplier No. 375540050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41019-45-8 | C11H6ClO3 | 221.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41019-45-8
Molecular Formula C11H6ClO3
Molecular Weight (g/mol) 221.62
MDL Number MFCD00216587
InChI Key XIPQHWUSDHTXOO-UHFFFAOYSA-M
PubChem CID 726693
SMILES [O-]C(=O)C1=CC=C(O1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 198.0°C to 201.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 5 g
Synonym 5-4-chlorophenyl-2-furoic acid, 5-4-chlorophenyl furan-2-carboxylic acid, 2-furancarboxylic acid, 5-4-chlorophenyl, 5-4-chloro-phenyl-furan-2-carboxylic acid, 2-furancarboxylic acid,5-4-chlorophenyl, acid, 62, acmc-1an70, 5-p-chlorophenyl furoic acid, 5-4-chlorophenyl furoic acid, 5-p-chlorophenyl-2furoic acid
Solubility Information Solubility in water: soluble. Other solubilities: ethanol,acetone,dmf,dioxane
IUPAC Name 5-(4-chlorophenyl)furan-2-carboxylate
Formula Weight 222.63
Percent Purity 95+%
Physical Form Powder
Chemical Name or Material 5-(4-Chlorophenyl)-2-furoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-(4-Chlorophenyl)-2-furoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 5-(4-Chlorophenyl)-2-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.