missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)-1,3,4-thiadiazole-2-thiol, 96%, Thermo Scientific™

Product Code. 15408895
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408895 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408895 Supplier Thermo Scientific Alfa Aesar Supplier No. H34342.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 205°C to 208°C
CAS 63857-85-2
Quantity 500 mg
Molecular Formula C8H5ClN2S2
MDL Number MFCD01569426
Synonym 5-4-chlorophenyl-1,3,4-thiadiazole-2-thiol, 5-4-chlorophenyl-3h-1,3,4-thiadiazole-2-thione, 2-mercapto-5-4'-chlorophenyl-1,3,4-thiadiazole, 5-4-chlorophenyl-2-mercapto-1,3,4-thiadiazole
InChI Key NGMTZWFKSLNRMT-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=NNC(=S)S2)Cl
IUPAC Name 5-(4-chlorophenyl)-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol) 228.712
PubChem CID 5129045
Formula Weight 228.72
Percent Purity 96%
Chemical Name or Material 5-(4-Chlorophenyl)-1,3,4-thiadiazole-2-thiol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.