missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol, 96%

Product Code. 15486237
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
15486237 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 15486237

Brand: Thermo Scientific Alfa Aesar H33429.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23766-28-1 | C8H5ClN2OS | 212.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 169°C to 170°C
CAS 23766-28-1
Quantity 5 g
Molecular Formula C8H5ClN2OS
MDL Number MFCD00227512
Synonym 5-4-chlorophenyl-1,3,4-oxadiazole-2-thiol, 5-4-chlorophenyl-1,3,4-oxadiazole-2 3h-thione, 1,3,4-oxadiazole-2-thiol, 5-4-chlorophenyl, 5-4-chlorophenyl-3h-1,3,4-oxadiazole-2-thione, 2-mercapto-5-4-chlorophenyl-1,3,4-oxadiazole, 5-4-chloro-phenyl-1,3,4 oxadiazole-2-thiol, acmc-20am8t, 5-4-chlorophenyl ;-1,3,4-oxadiazole-2 3h ;-thione, 5-4'-chlorophenyl-1,3,4-oxadiazole-2 3h-thione 11
InChI Key MUFWSGAENICYQA-UHFFFAOYSA-N
SMILES ClC1=CC=C(C=C1)C1=NNC(=S)O1
IUPAC Name 5-(4-chlorophenyl)-3H-1,3,4-oxadiazole-2-thione
Molecular Weight (g/mol) 212.65
PubChem CID 736579
Formula Weight 212.66
Percent Purity 96%
Chemical Name or Material 5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.