missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Bromophenyl)isoxazole, 98%, Thermo Scientific Chemicals

Product Code. 11469520
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469520 5 g 5g
11459520 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469520 Supplier Thermo Scientific Alfa Aesar Supplier No. H50342.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-(4-Bromophenyl)isoxazole is used as an active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(4-Bromophenyl)isoxazole is used as an active pharmaceutical ingredients.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7064-31-5
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD02183532
InChI Key AVMMSHZGFRWLRI-UHFFFAOYSA-N
Synonym 5-4-bromophenyl isoxazole, 5-4-bromophenyl-1,2-oxazole, acmc-209ofh
PubChem CID 2735608
IUPAC Name 5-(4-bromophenyl)-1,2-oxazole
SMILES C1=CC(=CC=C1C2=CC=NO2)Br

Specifications

Melting Point 115°C to 118°C
Quantity 5 g
Formula Weight 224.06
Percent Purity 98%
Chemical Name or Material 5-(4-Bromophenyl)isoxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.