missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Bromophenyl)isoxazole, 97%, Thermo Scientific™

Product Code. 10099181
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10099181 10 g Amber Glass Bottle 10g
10384512 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10099181 Supplier Thermo Scientific Maybridge Supplier No. CC12910EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7064-31-5
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD02183532
InChI Key AVMMSHZGFRWLRI-UHFFFAOYSA-N
Synonym 5-4-bromophenyl isoxazole, 5-4-bromophenyl-1,2-oxazole, acmc-209ofh
PubChem CID 2735608
IUPAC Name 5-(4-bromophenyl)-1,2-oxazole
SMILES C1=CC(=CC=C1C2=CC=NO2)Br

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 5-(4-Bromophenyl)isoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.