missing translation for 'onlineSavingsMsg'
Learn More

[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]-3-(2-pyridinyldithio)-L-alanine, TRC

Product Code. 30388967
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388967 100 mg 100mg
30405817 10 mg 10mg
30408666 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388967 Supplier Toronto Research Chemicals Supplier No. TRCH291770100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291770-37-1
Molecular Formula C18H24N4O4S3
Molecular Weight (g/mol) 456.6
IUPAC Name (2R)-2-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-3-(pyridin-2-yldisulfanyl)propanoic acid
SMILES OC(=O)[C@H](CSSc1ccccn1)NC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H24N4O4S3/c23-14(6-2-1-5-13-16-11(9-27-13)21-18(26)22-16)20-12(17(24)25)10-28-29-15-7-3-4-8-19-15/h3-4,7-8,11-13,16H,1-2,5-6,9-10H2,(H,20,23)(H,24,25)(H2,21,22,26)/t11-,12-,13-,16-/m0/s1
Formula Weight 456.096
Chemical Name or Material [5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]-3-(2-pyridinyldithio)-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.