missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-(3-Fluorophenyl)-1H-tetrazole, 97%
£31.24 - £92.52
Chemical Identifiers
CAS | 50907-20-5 |
---|---|
Molecular Formula | C7H5FN4 |
Molecular Weight (g/mol) | 164.143 |
MDL Number | MFCD01075702 |
InChI Key | SEYSTLGMSOLFBD-UHFFFAOYSA-N |
Synonym | 5-3-fluorophenyl-1h-tetrazole, 5-3-fluorophenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetrazole, 5-3-fluorophenyl-2h-1,2,3,4-tetrazole, 5-3-fluoro-phenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetraazole, acmc-20am9c, 3-fluorophenyl-tetrazole, 5-3-fluorophenyl tetrazole |
PubChem CID | 333943 |
IUPAC Name | 5-(3-fluorophenyl)-2H-tetrazole |
SMILES | C1=CC(=CC(=C1)F)C2=NNN=N2 |
Description
Chemical Identifiers
50907-20-5 | |
164.143 | |
SEYSTLGMSOLFBD-UHFFFAOYSA-N | |
333943 | |
C1=CC(=CC(=C1)F)C2=NNN=N2 |
C7H5FN4 | |
MFCD01075702 | |
5-3-fluorophenyl-1h-tetrazole, 5-3-fluorophenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetrazole, 5-3-fluorophenyl-2h-1,2,3,4-tetrazole, 5-3-fluoro-phenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetraazole, acmc-20am9c, 3-fluorophenyl-tetrazole, 5-3-fluorophenyl tetrazole | |
5-(3-fluorophenyl)-2H-tetrazole |
Specifications
50907-20-5 | |
C7H5FN4 | |
1104440 | |
SEYSTLGMSOLFBD-UHFFFAOYSA-N | |
5-(3-fluorophenyl)-2H-tetrazole | |
333943 | |
97% |
142°C to 144°C | |
MFCD01075702 | |
5-3-fluorophenyl-1h-tetrazole, 5-3-fluorophenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetrazole, 5-3-fluorophenyl-2h-1,2,3,4-tetrazole, 5-3-fluoro-phenyl-2h-tetrazole, 5-3-fluorophenyl-1h-1,2,3,4-tetraazole, acmc-20am9c, 3-fluorophenyl-tetrazole, 5-3-fluorophenyl tetrazole | |
C1=CC(=CC(=C1)F)C2=NNN=N2 | |
164.143 | |
164.14 | |
5-(3-Fluorophenyl)-1H-tetrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures