missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Chlorophenyl)-1H-tetrazole, 98+%

Product Code. 15448929
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15448929

Brand: Thermo Scientific Alfa Aesar L10699.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41421-28-7 | C7H4ClN4 | 179.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 139°C to 140°C
CAS 41421-28-7
Quantity 1 g
Molecular Formula C7H4ClN4
MDL Number MFCD00040951
Beilstein 140081
Synonym 5-3-chlorophenyl-1h-tetrazole, 5-3-chlorophenyl-2h-tetrazole, 5-3-chloro-phenyl-2h-tetrazole, 5-3-chlorophenyl-1h-1,2,3,4-tetrazole, 5-3-chlorophenyl-2h-1,2,3,4-tetrazole, 5-3-chlorophenyl-1h-1,2,3,4-tetraazole, 5-3-chlorophenyl tetraazole, 5-chlorophenyltetrazole, acmc-20am9f, 5-3-chlorophenyl tetrazole
InChI Key WKXXXGMIBYWPGE-UHFFFAOYSA-N
SMILES ClC1=CC=CC(=C1)C1=N[N-]N=N1
IUPAC Name 5-(3-chlorophenyl)-2H-tetrazole
Molecular Weight (g/mol) 179.59
PubChem CID 592520
Formula Weight 180.6
Percent Purity ≥98%
Chemical Name or Material 5-(3-Chlorophenyl)-1H-tetrazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.