missing translation for 'onlineSavingsMsg'
Learn More

5-[3-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. p-3540994
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10163314 250 mg Amber Glass Bottle 250mg
10045183 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163314 Supplier Thermo Scientific Maybridge Supplier No. CC49508CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-57-5
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
MDL Number MFCD09817470
InChI Key WKTARCMEMVHRLR-UHFFFAOYSA-N
Synonym 5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl bromide, 5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazol, alpha-bromo-3-3-methyl-1,2,4-oxadiazol-5-yl toluene, 1-bromomethyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzene, pubchem23332
PubChem CID 24229497
IUPAC Name 5-[3-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC(=CC=C2)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 5-[3-(Bromomethyl)phenyl]-3-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.