missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(3-((1H-1,2,4-Triazol-1-yl)methyl)-3-hydroxy-4,4-dimethylpentyl)-2-chlorophenol, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | 5-(3-((1H-1,2,4-Triazol-1-yl)methyl)-3-hydroxy-4,4-dimethylpentyl)-2-chlorophenol |
| Analyte or Component Names | 5-(3-((1H-1,2,4-Triazol-1-yl)methyl)-3-hydroxy-4,4-dimethylpentyl)-2-chlorophenol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16H22ClN3O2 |
| Synonym | Tebuconazole impurity |
| InChI Formula | InChI=1S/C16H22ClN3O2/c1-15(2,3)16(22,9-20-11-18-10-19-20)7-6-12-4-5-13(17)14(21)8-12/h4-5,8,10-11,21-22H,6-7,9H2,1-3H3 |
| Solution Type | Neat |
| SMILES | ClC1=CC=C(CCC(C(C)(C)C)(O)CN2C=NC=N2)C=C1O |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 323.82 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction