missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Thienyl)pentanoic acid, 95%

Product Code. 11397906
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397906 1 g 1g
11307916 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397906 Supplier Thermo Scientific Alfa Aesar Supplier No. L13122.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 21010-06-0 | C9H12O2S | 184.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21010-06-0
Molecular Formula C9H12O2S
Molecular Weight (g/mol) 184.25
MDL Number MFCD00151849
InChI Key OQFTZRHAQGXEQU-UHFFFAOYSA-N
Synonym 5-2-thienyl pentanoic acid, 5-thiophen-2-yl pentanoic acid, 2-thiophenepentanoic acid, 5-2-thienyl valeric acid, maybridge3_003072, acmc-1cq6n, 5-2-thienyl-pentanoic acid, 5-2-thienyl-n-valeric acid
PubChem CID 152324
IUPAC Name 5-thiophen-2-ylpentanoic acid
SMILES OC(=O)CCCCC1=CC=CS1

Specifications

Melting Point 39°C to 42°C
Quantity 1 g
Beilstein 126021
Formula Weight 184.26
Percent Purity 95%
Chemical Name or Material 5-(2-Thienyl)pentanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.