missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-thienyl)-1,3-oxazole, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Synonym | 5-2-thienyl-1,3-oxazole, 5-thiophen-2-yl oxazole, 5-2-thienyl oxazole, 5-thiophen-2-yl-1,3-oxazole, 5-thiophen-2-yl-oxazole, oxazole,5-2-thienyl, oxazole, 5-2-thienyl, 2-1,3-oxazol-5-yl thiophene |
Formula Weight | 151.19 |
Percent Purity | 95% |
Chemical Name or Material | 5-(2-Thienyl)-1, 3-oxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'