missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Thermo Scientific™

Product Code. 10178254
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10178254 5 g 5g
10049151 1 g 1g
10381615 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10178254 Supplier Thermo Scientific Maybridge Supplier No. CC18209DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-70-0
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
InChI Key CQCKNGVVCKWDLF-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methanol, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-4-thiazolyl thiophene-2-methanol, 5-2-methylthiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl thien-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methan-1-ol
PubChem CID 2776470
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
SMILES CC1=NC(=CS1)C2=CC=C(S2)CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 211.31
Percent Purity 95%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.