missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Hydroxyethyl)-4-methylthiazole, Mikromol™

Product Code. 30051344
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30051344 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30051344

Brand: Mikromol MM0049.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-(2-Hydroxyethyl)-4-methylthiazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(2-Hydroxyethyl)-4-methylthiazole
CAS 137-00-8
Analyte or Component Names 5-(2-Hydroxyethyl)-4-methylthiazole
Shipping Condition Room Temperature
Molecular Formula C6 H9 N O S
Certifications/Compliance ISO 17025
Synonym 5-(2-Hydroxyethyl)-4-methylthiazole, MHT
InChI Formula InChI=1S/C6H9NOS/c1-5-6(2-3-8)9-4-7-5/h4,8H,2-3H2,1H3
SMILES Cc1ncsc1CCO
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethanol
Molecular Weight (g/mol) 143.21
Quantity 100 mg
Formula Weight 143.0405 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.