missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)thiophene-2-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10103964
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10103964 1 g 1g
10295904 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103964 Supplier Thermo Scientific Maybridge Supplier No. CC47704DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-64-0
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.205
MDL Number MFCD06410178
InChI Key IXUVFAGSDDBEKJ-UHFFFAOYSA-N
Synonym 5-2-furyl thiophene-2-carbaldehyde, 5-furan-2-yl thiophene-2-carbaldehyde, 5-fur-2-yl thiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde,5-2-furanyl, 5-furan-2-yl thiophene-2-carboxaldehyde, 5-2-furanyl-2-thiophenecarboxaldehyde
PubChem CID 23004862
IUPAC Name 5-(furan-2-yl)thiophene-2-carbaldehyde
SMILES C1=COC(=C1)C2=CC=C(S2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 5-(2-Furyl)thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.