missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)pyridine-2-carbaldehyde, 95%, Thermo Scientific™

Product Code. 10452674
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10452674 250 mg Amber Glass Bottle 250mg
10565574 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10452674 Supplier Thermo Scientific Maybridge Supplier No. CC55104CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-45-2
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD10700053
InChI Key GUIKMKZMLLANPU-UHFFFAOYSA-N
Synonym 5-2-furyl pyridine-2-carbaldehyde, 5-furan-2-yl pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde,5-2-furanyl
PubChem CID 42555941
IUPAC Name 5-(furan-2-yl)pyridine-2-carbaldehyde
SMILES C1=COC(=C1)C2=CN=C(C=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.17
Percent Purity 95%
Chemical Name or Material 5-(2-Furyl)pyridine-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.