missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Furyl)-1H-pyrazol-3-yl]methanol, 97%, Thermo Scientific™

Product Code. 10005832
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10005832 1 g Amber Glass Bottle 1g
10585953 250 mg Amber Glass Bottle 250mg
10777105 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10005832 Supplier Thermo Scientific Maybridge Supplier No. CC35009DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 84978-67-6
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
InChI Key NBADTAIDQSLLHA-UHFFFAOYSA-N
Synonym 5-2-furyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1h-pyrazol-3-yl methanol, 3-furan-2-yl-1h-pyrazol-5-yl methanol, 5-2-furyl pyrazol-3-yl methan-1-ol, 1h-pyrazole-3-methanol,5-2-furanyl, 5-2-furyl-1h-pyrazol-4-yl methanol, 5-fur-2-yl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-2h-pyrazol-3-yl methanol
PubChem CID 7060534
IUPAC Name [3-(furan-2-yl)-1H-pyrazol-5-yl]methanol
SMILES C1=COC(=C1)C2=NNC(=C2)CO

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material [5-(2-Furyl)-1H-pyrazol-3-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.