missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10135832
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135832 1 g Amber Glass Bottle 1g
10709415 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10135832 Supplier Thermo Scientific Maybridge Supplier No. CC46902DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-47-1
Molecular Formula C9H7ClN2O2
Molecular Weight (g/mol) 210.617
MDL Number MFCD08271946
InChI Key RAYIOTXFHIKMIE-UHFFFAOYSA-N
Synonym 5-2-furyl-1-methyl-1h-pyrazole-3-carbonyl chloride, 5-furan-2-yl-1-methylpyrazole-3-carbonyl chloride, 5-furan-2-yl-1-methyl-1h-pyrazole-3-carbonyl chloride, 5-2-furyl-1-methylpyrazole-3-carbonyl chloride, 1h-pyrazole-3-carbonylchloride, 5-2-furanyl-1-methyl
PubChem CID 18525800
IUPAC Name 5-(furan-2-yl)-1-methylpyrazole-3-carbonyl chloride
SMILES CN1C(=CC(=N1)C(=O)Cl)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.62
Percent Purity 97%
Chemical Name or Material 5-(2-Furyl)-1-methyl-1H-pyrazole-3-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.