missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Fluorophenyl)-5-oxovaleric Acid, TRC

Product Code. 30401231
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401231 25 mg -
30392238 50 mg -
30404318 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401231 Supplier Toronto Research Chemicals Supplier No. TRCF48959525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199664-70-5
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.2
Synonym 2-Fluoro-delta-oxobenzenepentanoic Acid
SMILES OC(=O)CCCC(=O)c1ccccc1F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H11FO3/c12-9-5-2-1-4-8(9)10(13)6-3-7-11(14)15/h1-2,4-5H,3,6-7H2,(H,14,15)
Formula Weight 210.0692
Chemical Name or Material 5-(2-Fluorophenyl)-5-oxovaleric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.