missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Ethoxyphenyl)-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione, TRC

Product Code. 30373409
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373409 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373409 Supplier Toronto Research Chemicals Supplier No. TRCE89271025MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 479074-06-1
Molecular Formula C17H20N4OS
Quantity 25 mg
Synonym 5-(2-Ethoxyphenyl)-1,4-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione
InChI Formula InChI=1S/C17H20N4OS/c1-4-8-12-14-15(21(3)20-12)17(23)19-16(18-14)11-9-6-7-10-13(11)22-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,18,19,23)
SMILES CCCc1nn(C)c2C(=S)NC(=Nc12)c3ccccc3OCC
Molecular Weight (g/mol) 328.43
Formula Weight 328.1358
Chemical Name or Material 5-(2-Ethoxyphenyl)-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.