missing translation for 'onlineSavingsMsg'
Learn More

5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde, 95%, Thermo Scientific™

Product Code. 10365392
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10365392 1 g 1g
10417564 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10365392 Supplier Thermo Scientific Maybridge Supplier No. SEW06174DA
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-04-9
Molecular Formula C12H6ClF3O2
Molecular Weight (g/mol) 274.623
MDL Number MFCD01571128
InChI Key SAEPEPVLSICNRE-UHFFFAOYSA-N
Synonym 5-2-chloro-4-trifluoromethyl phenyl-2-furaldehyde, 5-2-chloro-4-trifluoromethyl phenyl furan-2-carbaldehyde, 2-furancarboxaldehyde,5-2-chloro-4-trifluoromethyl phenyl
PubChem CID 2781786
IUPAC Name 5-[2-chloro-4-(trifluoromethyl)phenyl]furan-2-carbaldehyde
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)C2=CC=C(O2)C=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 274.62
Percent Purity 95%
Chemical Name or Material 5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.