missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Catalog Number 10178794
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
375857-64-0
C9H7BrN2O
239.072
NYAFKUCVAVUCHW-UHFFFAOYSA-N
18001310
5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
CC1=NOC(=N1)C2=CC=CC=C2Br
This item is not returnable. View return policy
375857-64-0
C9H7BrN2O
239.072
NYAFKUCVAVUCHW-UHFFFAOYSA-N
18001310
5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
CC1=NOC(=N1)C2=CC=CC=C2Br
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 5-2-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-2-bromophenyl-3-methyl, 5-2-bromophenyl-3-methyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-2-bromophenyl-3-methyl
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 5-(2-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'