missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole, ≥97%, Thermo Scientific™

Catalog Number 10031974
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
906352-89-4
C11H8BrF3N2
305.098
MFCD09817512
UKDUPCWWWMHDRH-UHFFFAOYSA-N
5-2-bromophenyl-1-methyl-3-trifluoromethyl-1h-pyrazole, 5-2-bromophenyl-1-methyl-3-trifluoromethyl pyrazole
24229660
5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)pyrazole
CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2Br
This item is not returnable. View return policy
906352-89-4
C11H8BrF3N2
305.098
MFCD09817512
UKDUPCWWWMHDRH-UHFFFAOYSA-N
5-2-bromophenyl-1-methyl-3-trifluoromethyl-1h-pyrazole, 5-2-bromophenyl-1-methyl-3-trifluoromethyl pyrazole
24229660
5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)pyrazole
CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 305.1
Percent Purity 97%
Chemical Name or Material 5-(2-Bromophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'