missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-1,3-oxazole, ≥97%, Thermo Scientific™

Product Code. 10299712
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 328270-70-8
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD05668985
InChI Key JLTHLCLAPCIKJJ-UHFFFAOYSA-N
Synonym 5-2-bromophenyl oxazole, 5-2-bromophenyl-1,3-oxazole, oxazole,5-2-bromophenyl
PubChem CID 3780551
IUPAC Name 5-(2-bromophenyl)-1,3-oxazole
SMILES C1=CC=C(C(=C1)C2=CN=CO2)Br

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 5-(2-Bromophenyl)-1, 3-oxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'