missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromoethoxy)-2,3-dihydro-1,4-benzodioxine, 95%, Thermo Scientific™

Product Code. 10782303
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 1710-62-9
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H11BrO3
MDL Number MFCD00099411
Quantity 1 g
Synonym 5-2-bromoethoxy-2,3-dihydro-1,4-benzodioxine, 5-2-bromoethoxy-2,3-dihydro-1,4-benzodioxin, 5-2-bromoethoxy-2,3-dihydrobenzo b 1,4 dioxine, 5-2-bromoethoxy-1,4-benzodioxane, 1,4-benzodioxin,5-2-bromoethoxy-2,3-dihydro, 1,4-benzodioxin, 5-2-bromoethoxy-2,3-dihydro, 5-2-bromoethoxy-2,3-dihydro-benzo 1,4 dioxine, 1-2h,3h-benzo 2,3-e 1,4-dioxin-5-yloxy-2-bromoethane
InChI Key IHXJWHBJQFUJBZ-UHFFFAOYSA-N
SMILES BrCCOC1=CC=CC2=C1OCCO2
IUPAC Name 5-(2-bromoethoxy)-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol) 259.10
PubChem CID 2735566
Formula Weight 259.1
Percent Purity 95%
Chemical Name or Material 5-(2-Bromoethoxy)-2, 3-dihydro-1, 4-benzodioxine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'