missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Aminoethylamino)-1-Naphthalene sulfonic acid, MP Biomedicals™

Product Code. 11418841 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418841 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11418841 Supplier MP Biomedicals Supplier No. 0215474001
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Synthetic. Soluble in DMSO and 1N NaOH (2.0mg/mL). Free Acid

TRUSTED_SUSTAINABILITY

Specifications

CAS 50402-56-7
Color Gray-Yellow
Molecular Formula C12H14N2O3S
MDL Number MFCD00066932
Quantity 1 g
Synonym 5-2-aminoethyl amino naphthalene-1-sulfonic acid, 1,5-edans, 5-2-aminoethylamino-1-naphthalenesulfonic acid, edans, 5-2-aminoethylamino-1-naphthalene sulfonic acid, 5-2-aminoethylamino naphthalene-1-sulfonic acid, n-aminoethyl-5-naphthylamine-1-sulfonic acid, 1-naphthalenesulfonicacid, 5-2-aminoethyl amino
InChI Key SJQRQOKXQKVJGJ-UHFFFAOYSA-N
SMILES NCCNC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O
IUPAC Name 5-[(2-aminoethyl)amino]naphthalene-1-sulfonic acid
Molecular Weight (g/mol) 266.32
PubChem CID 92329
Formula Weight 266.32
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Aminoethylamino)-1-Naphthalene sulfonic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.