missing translation for 'onlineSavingsMsg'
Learn More

5-[(2,5-Dimethyl-1-phenyl-1H-pyrrol-3-yl)methylene]-2-thioxo-4-thiazolidinone, TRC

Product Code. 30380491
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380491 50 mg -
30372442 250 mg -
30381745 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380491 Supplier Toronto Research Chemicals Supplier No. TRCD59625050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312716-40-8
Molecular Formula C16H14N2OS2
Molecular Weight (g/mol) 314.43
Synonym Optovin Analog 6b8
SMILES Cc1cc(\C=C/2\SC(=S)NC2=O)c(C)n1c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H14N2OS2/c1-10-8-12(9-14-15(19)17-16(20)21-14)11(2)18(10)13-6-4-3-5-7-13/h3-9H,1-2H3,(H,17,19,20)/b14-9+
Formula Weight 314.0548
Chemical Name or Material 5-[(2,5-Dimethyl-1-phenyl-1H-pyrrol-3-yl)methylene]-2-thioxo-4-thiazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.