missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5,15-Diacetyl-3-benzoyllathyrol

Product Code. 16441438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441438 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441438 Supplier MedChemExpress Supplier No. HYN0562
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5,15-Diacetyl-3-benzoyllathyrol is one of the lathyrane diterpenoids, that has anti-cancer activity.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 522.63 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 218916-52-0
Product Type Natural Products
Purity 98.85%
Solubility DMSO : 100 mg/mL (191.34 mM∣Need ultrasonic)
Synonym Euphorbia factor L3
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.