missing translation for 'onlineSavingsMsg'
Learn More

5,12-Napthacenequinone, TRC

Product Code. 30483042
Change view
Click to view available options
Quantity:
2.5 g
5 g
Unit Size:
2.5g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483042 5 g 5g
30472008 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483042 Supplier Toronto Research Chemicals Supplier No. TRCN3440005G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,12-Napthacenequinone
CAS 1090-13-7
Molecular Formula C18H10O2
Quantity 5 g
Synonym Naphthacenequinone, 5,12-Naphthacenequinone, 5,12-Tetracenequinone, Benz[b]anthracene-5,12-dione, NSC 401183, Naphthacene-6,11-quinone, Tetracenemonoquinone
InChI Formula InChI=1S/C18H10O2/c19-17-13-7-3-4-8-14(13)18(20)16-10-12-6-2-1-5-11(12)9-15(16)17/h1-10H
SMILES O=C1c2ccccc2C(=O)c3cc4ccccc4cc13
IUPAC Name tetracene-5,12-dione
Molecular Weight (g/mol) 258.27
Formula Weight 258.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.