missing translation for 'onlineSavingsMsg'
Learn More

5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals

Product Code. 15419165
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419165 5 g 5g
15409165 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419165 Supplier Thermo Scientific Alfa Aesar Supplier No. H51117.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1218790-44-3 | Molecular Formula: C16H26BNO2S | Molecular Weight: 307.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218790-44-3
Molecular Formula C16H26BNO2S
Molecular Weight (g/mol) 307.259
MDL Number MFCD11113036
InChI Key KDPVLCCGXRCQCV-UHFFFAOYSA-N
Synonym 5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
PubChem CID 46739745
IUPAC Name 1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]piperidine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3

Specifications

Quantity 5 g
Formula Weight 307.26
Chemical Name or Material 5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.