Description
- Tiva Washer Accessory
- 4th Dosing Pump
- Dos. 4 Disinfectant/additive pump with flowmeter, for proper product dosage
Specifications
Specifications
| Material | Plastic |
| Product Type | Pump |
| For Use With (Equipment) | Tiva Washer |
| Quantity | 1 Ea. |