missing translation for 'onlineSavingsMsg'
Learn More

(4S3-[(2R,5S)-5-(4-Fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone, TRC

Product Code. 30135830
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135830 250 mg 250mg
30009610 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135830 Supplier Toronto Research Chemicals Supplier No. TRCF598445250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 272778-12-8
Molecular Formula C39 H46 F2 N2 O5 Si2
Molecular Weight (g/mol) 716.961
Synonym (4S3-[(2R,5S)-5-(4-Fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone, 2-Oxazolidinone, 3-[(2R,5S)-5-(4-fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-, (4S)- (9CI, ACI), (4S)-3-[(2R,5S)-5-(4-Fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(tri
IUPAC Name (4S)-3-[(2R,5S)-2-[(S)-(4-fluoroanilino)-(4-trimethylsilyloxyphenyl)methyl]-5-(4-fluorophenyl)-5-trimethylsilyloxypentanoyl]-4-phenyl-1,3-oxazolidin-2-one
SMILES O=C1OCC(C=2C=CC=CC2)N1C(=O)C(CCC(O[Si](C)(C)C)C3=CC=C(F)C=C3)C(NC4=CC=C(F)C=C4)C5=CC=C(O[Si](C)(C)C)C=C5

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C39H46F2N2O5Si2/c1-49(2,3)47-33-22-14-29(15-23-33)37(42-32-20-18-31(41)19-21-32)34(24-25-36(48-50(4,5)6)28-12-16-30(40)17-13-28)38(44)43-35(26-46-39(43)45)27-10-8-7-9-11-27/h7-23,34-37,42H,24-26H2,1-6H3/t34-,35-,36+,37-/m1/s1
Formula Weight 716.291
Percent Purity >95%
Chemical Name or Material (4S)-3-[(2R,5S)-2-[(S)-(4-fluoroanilino)-(4-trimethylsilyloxyphenyl)methyl]-5-(4-fluorophenyl)-5-trimethylsilyloxypentanoyl]-4-phenyl-1,3-oxazolidin-2-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.