missing translation for 'onlineSavingsMsg'
Learn More

(4S)-6-Chloro-2-(4-methoxyphenyl)-4-(pent-3-en-1-yn-1-yl)-4-(trifluoromethyl)-2,4-dihydro-1H-benzo[d][1,3]oxazine, TRC

Product Code. 30354852
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354852 5 mg 5mg
30364491 100 mg -
30357704 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354852 Supplier Toronto Research Chemicals Supplier No. TRCC3804055MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21 H17 Cl F3 N O2
Molecular Weight (g/mol) 407.813
IUPAC Name (4S)-6-chloro-2-(4-methoxyphenyl)-4-[(E)-pent-3-en-1-ynyl]-4-(trifluoromethyl)-1,2-dihydro-3,1-benzoxazine
SMILES COc1ccc(cc1)C2Nc3ccc(Cl)cc3[C@@](O2)(C#C\C=C\C)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H17ClF3NO2/c1-3-4-5-12-20(21(23,24)25)17-13-15(22)8-11-18(17)26-19(28-20)14-6-9-16(27-2)10-7-14/h3-4,6-11,13,19,26H,1-2H3/b4-3+/t19?,20-/m0/s1
Formula Weight 407.09
Chemical Name or Material (4S)-6-Chloro-2-(4-methoxyphenyl)-4-(pent-3-en-1-yn-1-yl)-4-(trifluoromethyl)-2,4-dihydro-1H-benzo[d][1,3]oxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.